2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC49432-1G PC49432-250MG
76605-890EA 377.53 USD
76605-890 76604-048
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31759997
CAS Number: 1800423-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR