2-Bromo-3-(trifluoromethyl)benzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [2-Bromo-3-(trifluoromethyl)phenyl]methanol, (2-Bromo-3-(trifluoromethyl)phenyl)methanol

PC49387-1G PC49387-5G
76606-226EA 389.21 USD
76606-226 76605-244
2-Bromo-3-(trifluoromethyl)benzyl alcohol 95%
2-Bromo-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185396
CAS Number: 1214330-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR