2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2,2-Trifluoro-1-(2-(trifluoromethyl)phenyl)ethanone, 2,2,2-Trifluoro-1-[2-(trifluoromethyl)phenyl]ethan-1-one, 2-(Trifluoroacetyl)benzotrifluoride

PC49384-1G PC49384-5G
76606-950EA 94 USD
76606-950 76607-642
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone 96%
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₄F₆O
MW: 242.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380778
CAS Number: 42452-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR