2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Iodo-6-nitro-4-[(trifluoromethyl)sulphanyl]phenol, 4-Hydroxy-3-iodo-5-nitrophenyl trifluoromethyl sulfide, 2-Iodo-6-nitro-4-[(trifluoromethyl)sulfanyl]phenol, 4-Hydroxy-3-iodo-5-nitrophenyl trifluoromethyl sulphide

PC49360-1G PC49360-250MG
76607-032EA 705.22 USD
76607-032 76605-016
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol
Formula: C₇H₃F₃INO₃S
MW: 365.068 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24850032
CAS Number: 1440535-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR