4-Bromo-2-(trifluoromethyl)benzeneboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-2-(trifluoromethyl)phenylboronic acid, 2-Borono-5-bromobenzotrifluoride, (4-Bromo-2-(trifluoromethyl)phenyl)boronic acid

PC49292-1G PC49292-5G
76605-410EA 59.06 USD
76605-410 76607-566
4-Bromo-2-(trifluoromethyl)benzeneboronic acid
4-Bromo-2-(trifluoromethyl)benzeneboronic acid
Formula: C₇H₅BBrF₃O₂
MW: 268.824 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18447612
CAS Number: 1394346-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR