3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Methyl-7-(trifluoromethyl)-1H-indazol-3-amine

PC49029-1G PC49029-250MG
76605-038EA 339.78 USD
76605-038 76604-872
3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole
3-Amino-1-methyl-7-(trifluoromethyl)-1H-indazole
Formula: C₉H₈F₃N₃
MW: 215.175 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD11847095
CAS Number: 1260678-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR