5-Bromo-2,3,4-trifluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC48807-250MG PC48807-5G PC48807-1G
76618-408EA 20.26 USD
76618-408 76618-410 76618-406
5-Bromo-2,3,4-trifluorobenzonitrile 95%
5-Bromo-2,3,4-trifluorobenzonitrile
Formula: C₇HBrF₃N
Storage Temperature: Ambient
MDL Number: MFCD26679060
CAS Number: 1503508-74-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR