3,3,3-Trifluoropropane-1-sulfonamide 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC49037-1G PC49037-250MG
76604-702EA 254.68 USD
76604-702 76607-260
3,3,3-Trifluoropropane-1-sulfonamide 95%
3,3,3-Trifluoropropane-1-sulfonamide
Formula: C₃H₆F₃NO₂S
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD08704575
CAS Number: 1033906-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR