3-Amino-5-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC49080-1G PC49080-250MG PC49080-5G
76606-990EA 26.48 USD
76606-990 76605-586 76607-860
3-Amino-5-(trifluoromethyl)benzonitrile 98%
3-Amino-5-(trifluoromethyl)benzonitrile
Formula: C8H5F3N2
MW: 186.1339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09955455
CAS Number: 49674-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR