3-Bromo-2,6-difluorobenzoic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

PC49617-1G PC49617-25G PC49617-5G
76606-470EA 23.44 USD
76606-470 76605-774 76604-996
3-Bromo-2,6-difluorobenzoic acid 96%
3-Bromo-2,6-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 237.0000 g/mol
Melting Pt: 140…141 °C
Storage Temperature: Ambient
MDL Number: MFCD09261248
CAS Number: 28314-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR