2-Fluoro-5-hydroxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC49096-5G PC49096-1G
76605-360EA 161.8 USD
76605-360 76604-528
2-Fluoro-5-hydroxybenzaldehyde
2-Fluoro-5-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.1100 g/mol
Melting Pt: 74…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038469
CAS Number: 103438-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR