N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Decyldimethylazaniumyl)propane-1-sulphonate3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulfonate3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate3-(Decyldimethylazaniumyl)propane-1-sulfonate

45968-0050 45968-0250 45968-1000
TS45968-0050EA 58.7 USD
TS45968-0050 TS45968-0250 TS45968-1000
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10) 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10)
Formula: C₁₅H₃₃NO₃S
MW: 307.4924 g/mol
Melting Pt: 229 °C
Storage Temperature: Ambient
MDL Number: MFCD00036908
CAS Number: 15163-36-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 227 to 233 °C
Loss on drying ≤2 %
Solubility (10% in water) Clear colorless solution
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR