1-Boc-4-hydroxypiperidine 98+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-BOC-4-HydroxypiperidineN-Boc-4-piperidinoltert-butyl 4-hydroxypiperidine-1-carboxylatetert-Butyl 4-hydroxypiperidine-1-carboxylate

OR5404-25G OR5404-500G OR5404-100G
76591-734EA 16.48 USD
76591-734 76591-736 76591-738
1-Boc-4-hydroxypiperidine 98+%
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR