4-Formylphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-Formylphenyl)metaboric acid4-Formylbenzeneboronic acid4-BoronobenzaldehydeDihydroxy(4-formylphenyl)borane

OR6999-25G OR6999-5G OR6999-100G
76587-336EA 13.8 USD
76587-336 76587-338 76587-340
4-Formylphenylboronic acid
4-Formylphenylboronic acid
Formula: C₇H₇BO₃
MW: 149.94 g/mol
Melting Pt: 237…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00151823
CAS Number: 87199-17-5
EINECS: 438-670-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR