Universal U-Electrode, Mettler-Toledo
Supplier: Mettler-Toledo
11107764
76580-208EA 4235 USD
76580-208
Universal U-Electrode, Mettler-Toledo
Electrodes Other Electrodes
Anti-static solution for samples and tare containers with dimensions of 12×25 cm.
  • The U-electrode can be placed next to the balance, and will remove the static charge to avoid inaccurate weighing results