2-Chloro-4-iodopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

37842-0050 37842-0250
TS37842-0250EA 165 USD
TS37842-0250 TS37842-0050
2-Chloro-4-iodopyridine 98%
2-Chloro-4-iodopyridine
Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD01861983
CAS Number: 153034-86-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown-yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 45 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR