1,6-Diphenyl-1,3,5-hexatriene 98%

Supplier: Thermo Scientific Chemicals
Warning

11730-0010 11730-0050
TS11730-0010EA 68.3 USD
TS11730-0010 TS11730-0050
1,6-Diphenyl-1,3,5-hexatriene 98%
1,6-Diphenyl-1,3,5-hexatriene
Formula: (C₆H₅)CHCHCHCHCHCH(C₆H₅)
MW: 232.33 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 199…203 °C
MDL Number: MFCD00004793
CAS Number: 1720-32-7
EINECS: 217-011-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥97.5 %
Bromide (Br) ≤50 ppm
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Free chlorine ≤1 ppm (as Cl)
Ammonium (NH4) ≤3 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (ICP-OES) ≤1 ppm
Extractable organic subst (about 5 ppm) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR