Formononetin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one7-Hydroxy-3-(4-methoxyphenyl)chromone7-Hydroxy-4'-methoxyisoflavone

34248-0010
TS34248-0010EA 159 USD
TS34248-0010
Formononetin 98%
Formononetin
Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00016948
CAS Number: 485-72-3

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 256 to 261 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR