Stigmasterol 95%

Supplier: Thermo Scientific Chemicals

13276-0250 13276-0050
TS13276-0050EA 179 USD
TS13276-0050 TS13276-0250
Stigmasterol 95%
Stigmasterol
Formula: C₂₉H₄₈O
Melting Pt: 161…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003630
CAS Number: 83-48-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 161 to 170 °C
Loss on drying ≤1 %
GC ≥94 %
Specific optical rotation -48° to -52° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR