2-Ethoxybenzamide 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethenzamide

11787-0050 11787-1000
TS11787-0050EA 23.5 USD
TS11787-0050 TS11787-1000
2-Ethoxybenzamide 97%
2-Ethoxybenzamide
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 129…134 °C
MDL Number: MFCD00007977
CAS Number: 938-73-8
EINECS: 213-346-4

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 129 to 134 °C
HPLC ≥96.0 %
Ferro compounds ≤0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR