Eriochrome cyanine R, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Chromoxane Cyanine R

21515-0050
TS21515-0050EA 25 USD
TS21515-0050
Eriochrome cyanine R, pure
Eriochrome cyanine R
Formula: C₂₃H₁₅Na₃O₉S
Storage Temperature: Ambient
MDL Number: MFCD00012411
CAS Number: 3564-18-9
EINECS: 222-641-7

Order Now

Specification Test Results

Appearance Red to dark brown Powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR