Mercury(II) chloride 99.5+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercury dichloride
, Mercuric chloride
, Mercury Chloride
AC42393-5000
AC42393-0025
TS42393-5000EA
276
USD
TS42393-5000
TS42393-0025
Mercury(II) chloride 99.5+% ACS
Mercury (II) chloride
Formula:
HgCl₂ MW: 271.5 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 277 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011041 CAS Number: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1,II |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration Complexometric | ≥99.5 % |
Solubility | (in ethyl ether) Passes test |
Iron (Fe) | ≤20 ppm |
Residue after reduction | ≤0.02 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...