Calcein AM ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Calcein-AMCalcein O,O′-diacetate tetrakis(acetoxymethyl) esterCalcein acetoxymethyl esterTetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

22002
76483-464EA 124.31 USD
76483-464
Calcein AM ≥90%
Calcein AM

Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein.

Formula: C₄₆H₄₆N₂O₂₃
MW: 994.86 g/mol
MDL Number: MFCD05861516
CAS Number: 148504-34-1
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR