Zinquin

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Zinquin A

21260
76483-376EA 124.31 USD
76483-376
Zinquin
Zinquin

Zinquin is a zinc-sensitive, cell-impermeable fluorescent probe.

Formula: C₁₉H₁₈N₂O₅S
MW: 386.42 g/mol
MDL Number: MFCD03452711
CAS Number: 151606-29-0
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR