BAPTA-AM ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) EsterBAPTA-AM esterTetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate

21001
76483-116EA 96.26 USD
76483-116
BAPTA-AM ≥95%
BAPTA-AM

BAPTA AM is cell-permeable version of BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid), a cell-permeable calcium chelator.

Formula: C₃₄H₄₀N₂O₁₈
MW: 764.69 g/mol
MDL Number: MFCD00036696
CAS Number: 126150-97-8
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR