7-Aminoactinomycin D (7-AAD) ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: NSC-2397597-AADActinomy2,7-diamino-4,6-dimethyl-3-oxo-1-N,9-N-bis[7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]phenoxazine-1,9-dicarboxamide

17501
76482-838EA 96.26 USD
76482-838
7-Aminoactinomycin D (7-AAD) ≥95%
7-Aminoactinomycin D (7-AAD)

7-Amino actinomycin D (7-AAD) is a non-permeant dye that can be used to identify non-viable cells.

Formula: C₆₂H₈₇N₁₃O₁₆
MW: 1270.43 g/mol
MDL Number: MFCD00077295
CAS Number: 7240-37-1
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR