PREempt® Plus Disinfectant Wipes, Contec
Supplier: Contec
2B221
76499-144CS 268.13 USD
76499-144
PREempt® Plus Disinfectant Wipes, Contec
Wipes Disposable Wipes and Towels Disinfectant Wipes
One-step disinfectant cleaner wipes utilizing accelerated Hydrogen Peroxide®.
  • Faster kill times make it easier to maintain compliance and appropriate dwell time
  • Less residue makes it more suitable for critical environments
  • Presaturated wipes leave appropriate disinfectant on surfaces
  • Contains low levels of hydrogen peroxide and nonhazardous, allowing for cost-effective disposal
  • Lowest EPA toxicity rating requires no PPE, contains no VOCs, and is odor-free for better operator acceptability
  • Compatible with a wide range of materials for less wear on surfaces and equipment
PREempt® Plus is a new and improved formulation of PREempt RTU that features faster kill times and lower residue. PREempt Plus disinfectant wipes are presaturated utilizing Accelerated Hydrogen Peroxide® (AHP®), a patented synergistic blend of commonly used ingredients, that when combined with low levels of hydrogen peroxide dramatically increasing its efficacy and cleaning performance.
These disinfectant wipes kill viruses in 30 seconds, bacteria in 1 minute, fungi in 30 seconds to 1 minute and TB in 1 minute.
Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and laboratory areas including workstations, fume hoods, equipment and other hard non-porous environmental surfaces.
Packaging: Wipes are packaged 160 sheets per cannister, 12 cannisters per case.