Formononetin 98+%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one7-Hydroxy-3-(4-methoxyphenyl)chromone7-Hydroxy-4'-methoxyisoflavone

A204811-100MG A204811-100G A204811-5G A204811-25G A204811-1G A204811-10G
77030-268EA 107.16 USD
77030-268 77030-270 77030-272 103817-570 77030-274 77600-670
Formononetin 98+%
Formononetin

Caution: for Research Use Only

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
Boiling Pt: 479.4 °C (760 mm Hg)
Melting Pt: 256…260 °C
Flash Pt: 183.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00016948
CAS Number: 485-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR