Polyethylene glycol 400 viscous liquid
Supplier: Electron Microscopy Sciences
Synonyms:
PEG 400
19720
100504-208EA
60.97
USD
100504-208
Polyethylene glycol 400 viscous liquid
Polyethylene glycol 400
An embedding medium for microscopy and histochemistry
Formula:
H(OCH₂CH₂)nOH MW: 400 g/mol Density: 1.1254 (20 °C) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...