NanoXact Silver Nanoplates 1 mg/ml PVP Surface (Polyvinylpyrrolidone), nanoComposix
Supplier: nanoComposix
SPPH660-1M SPPH800-5M SPPH1064-1M SPPH1064-10M SPPH800-10M SPPH980-10M SPPH1064-5M SPPH660-10M SPPH660-5M SPPH800-30M SPPH1064-30M SPPH980-5M SPPH800-1M SPPH980-1M SPPH980-30M SPPH660-30M
103820-850EA 1130 USD
103820-850 103820-840 103820-830 103820-842 103820-820 103820-832 103820-822 103820-844 103820-834 103820-824 103820-846 103820-836 103820-826 103820-848 103820-838 103820-828
NanoXact Silver Nanoplates 1 mg/ml PVP Surface (Polyvinylpyrrolidone), nanoComposix
Beads Microcarriers
High concentration silver nanoplates (nanodiscs) with peak absorbances from 660 to 1064 nm wavelengths.
  • Silver nanoplates
  • High quality nanoparticles, precisely engineered and highly characterized
  • Ultra strong light absorbing and scattering materials (OD's of ~100 to 160)
  • High concentration 1 mg/ml nanomaterial
Platelet silver nanoparticles (a.k.a. silver nanoplates or silver nanodisks). Peak surface plasmon resonances (SPR) with peak absorbances across wavelengths from 660 to 1064 nm. Provided at 1 mg/ml high concentration. PVP surface provides strong steric stability (40 kD PVP). Precisely engineered, unagglomerated nanoplates. Ultra to strong light absorbing and scattering materials.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.