Cyanine 647NS succinimidyl ester cyanine dye, non-sulfonated
Supplier: Biotium
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS, and Cyanine 647NS are designed to match by charge and molecular weight for 2D DIGE platform.
- Ex/Em 639/668 nm
- MW added to protein: 439
- Dark blue solid soluble in DMF or DMSO
- Store desiccated at -20°C; protect from light
Cyanine NS succinimidyl esters are non-sulfonated cyanine dyes that can be used to label proteins, oligonucleotides and nucleic acids. Available in 3 colors, Cyanine 488NS, Cyanine 555NS and Cyanine 647NS are non-sulfonated versions of Cy®2, Cy®3, and Cy®5, respectively. They are designed to match by charge and molecular weight for 2D DIGE platform. Biotium offers these dyes as high quality amine-reactive succinimidyl esters.
Also see sulfonated Cyanine Dye Succinimidyl Esters and our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy2, Cy3, Cy5 are registered trademarks of GE Healthcare.