Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Hexahydropyridine
, Hexazane
, Pentamethyleneimine
80640-500ML
80640-1L
BJ80640-500MLEA
224.37
USD
BJ80640-500ML
BJ80640-1L
Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™
Piperidine
Beilstein Number: 102438
RTECS Number: TM3500000
UNSPSC Code: 12190000
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 10.3 %(V) Melting Pt: -10 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005979 CAS Number: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8,I |
Specification Test Results
Refractive Index | n20/D 1.452 (lit.) |
Identity (IR) | complying |
Assay (GC) | Min. 99.5 % |
Refractive index (n 20/D) | 1.452 - 1.454 |
Non-volatile matter | Max. 0.02 % |
Water (Karl Fischer) | Max. 0.3 % |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Tin (Sn) | Max. 0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...