Mercury(II) chloride ≥99.5% (dried basis), Puriss. p.a. ACS, ISO, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Mercury dichloride
, Mercuric chloride
, Mercury Chloride
31005-500G
31005-100G
BJ31005-500GEA
592.93
USD
BJ31005-500G
BJ31005-100G
Mercury(II) chloride ≥99.5% (dried basis), Puriss. p.a. ACS, ISO, Fluka™
Mercury (II) chloride
Formula:
HgCl₂ MW: 271.5 g/mol Boiling Pt: 302 °C Melting Pt: 276 °C Density: 5.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011041 CAS Number: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1,II |
Specification Test Results
Assay (calculated to the dried substance) | Min. 99.5 % |
Loss on drying (on P₂O₅) | Max. 1.0 % |
Residue on ignition a. reducing with HCOOH (SO₄)* | Max. 0.02 % |
Water insoluble matter | Max. 0.01 % |
Silver (Ag) | Max. 0.0005 % |
Calcium (Ca) | Max. 0.001 % |
Cadmium (Cd) | Max. 0.0005 % |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 0.001 % |
Iron (Fe) | Max. 0.0005 % |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.001 % |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 0.005 % |
Nickel (Ni) | Max. 0.0005 % |
Lead (Pb) | Max. 0.001 % |
Zinc (Zn) | Max. 0.0005 % |
Other heavy metals (as Pb) | Max. 0.001 % |
Mercury(I) chloride | Max. 0.05 % |
Total S (as SO₄) | Max. 50 ppm |
Sulfate (SO₄) | Max. 0.02 % |
Total N | Max. 0.002 % |
In ether insoluble matter | complying |
Appearance of the solution | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...