Catechol violet indicator for metal titration, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Pyrocatechol Violet
32672-5G
32672-1G
BJ32672-5GEA
112.5
USD
BJ32672-5G
BJ32672-1G
Catechol violet indicator for metal titration, Fluka™
Catechol violet
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Beilstein Number: 353938
Formula:
C₁₉H₁₄O₇S Melting Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005868 CAS Number: 115-41-3 EINECS: 204-088-3 |
Specification Test Results
Change range | complying |
Indicator Solution 0,1 % H₂O | complying |
Suitable for metal definition | complying |
Absorption maximum | 436 - 446 nm |
Absorbtion (1%/1cm) | 400 - 500 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...