6-CFDA (6-Carboxyfluorescein diacetate, single isomer) ≥99% (by TLC) fluorescent pH indicator

Supplier: Biotium

Synonyms: 6-CFDA6-Carboxyfluorescein diacetate6-Carboxy-di-O-acetylfluorescein

51021
89139-264EA 176.71 USD
89139-264
6-CFDA (6-Carboxyfluorescein diacetate, single isomer) ≥99% (by TLC) fluorescent pH indicator
6-CFDA (6-Carboxyfluorescein diacetate)

6-CFDA is membrane-permeant and thus can be loaded into cells via incubation. Once inside cells, 6-CFDA is hydrolyzed by intracellular esterases to 6-carboxyfluorescein.


  • Cell permeability: Membrane permeant
  • Colors: Green
  • λex\λem 492nm\517nm (pH 9, after hydrolysis)
  • Extinction Coefficient (ε) 78,000 (pH 9, after hydrolysis)
  • Off-white solid soluble in DMSO
  • Store desiccated at -20°C

Formula: C₂₅H₁₆O₉
MW: 460.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036871
CAS Number: 3348-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR