Crystal violet 1% lilies solution

Supplier: Electron Microscopy Sciences

Synonyms: Basic Violet 3GentiavermMethylrosanilide chlorideHexamethyl VioletVianinGentian violetMethyl violet 10BCI 42555Hexamethyl-p-rosaniline chlorideN-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chlorideDimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchloridViocid

26088-17
101410-996EA 58.34 USD
101410-996
Crystal violet 1% lilies solution
Crystal Violet

Crystal Violet solutions are prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology


Available in a concentrations of 1.0% aqueous or Huckers solution, Lieb's stock solution, or Stirling's solution.

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
https://us.vwr.com/stibo/web/std.lang.all/48/02/28984802.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR