Piperidine ≥99% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: HexahydropyridineHexazanePentamethyleneimine

Management of Change (MOC) category = N

BAKR2895-05 BAKR2895-05 BAKR2895-98 BAKR2895-98
JT2895-05CS 497.36 USD
JT2895-05 CAJT2895-98 JT2895-98 CAJT2895-5
Piperidine ≥99% (by GC), J.T.Baker®
Piperidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Density: 0.86 g/cm³
Flash Pt: 16.00 °C
MDL Number: MFCD00005979
CAS Number: 110-89-4
UN: 2401
ADR: 8,I

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₅H₁₁N) (by GC) ≥99 %
Water (H₂O) ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR