Starter Assembly for Arlink® 8000 Workstations, Arlink
Supplier: Arlink
ARL-8897 ARL-8694
76240-246EA 0.73 USD
76240-246 76268-292
Starter Assembly for Arlink® 8000 Workstations, Arlink
Furniture Tables and Workbenches
The starter assembly is the first component for each workstation and includes two columns with outer and inner trim, and a closed stabilizer panel. Optional open stabilizer panel may be ordered for full access from the rear of the workstation.