TMR Biocytin

Supplier: Anaspec Inc

Synonyms: Biocytin conjugates5-(&-6)-Tetramethylrhodamine Biocytin

AS-60658
103003-308EA 249.92 USD
103003-308
TMR Biocytin
Biocytin TMR

Orange fluorescent biotin used for studying avidin binding


  • Spectral properties: Abs/Em = 554/581 nm
  • Solvent system: DMF or DMSO
  • Appearance: solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR