2,3-Naphthalenedialdehyde (NDA) ≥95% (by HPLC), Ultra Pure Grade
Supplier: Anaspec Inc
Synonyms:
Naphthalene-2,3-dicarboxaldehyde
, Naphthalene-2,3-dicarbaldehyde
, NDA
AS-83010
103011-112EA
226.54
USD
103011-112
2,3-Naphthalenedialdehyde (NDA) ≥95% (by HPLC), Ultra Pure Grade
2,3-Naphthalenedialdehyde (NDA)
Fluorogenic indicator for primary amino group; Widely used for quantitation of peptides and proteins.
Formula:
C₁₂H₈O₂ MW: 184.19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00016618 CAS Number: 7149-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...