Anti-FOLR1 Mouse Monoclonal Antibody [clone: 548908]

Supplier: Enzo Life Sciences
ABS378-0100
76203-732EA 718 USD
76203-732
Anti-FOLR1 Mouse Monoclonal Antibody [clone: 548908]
Antibodies
Recommended Applications: ELISA, Flow Cytometry, ICC, WB

Type: Primary
Antigen: FOLR1
Clonality: Monoclonal
Clone: 548908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR