4-Chloromercuribenzoic acid, white solid

Supplier: TORONTO RESEARCH CHEMICALS MS
Danger

C3678001G
100579-796EA 583.08 USD
100579-796
4-Chloromercuribenzoic acid, white solid
4-Chloromercuriobenzoic acid

4-Chloromercuribenzoic acid is a useful sulfhydryl reagent that reacts with histidine 225C, a residue of the NhaA-Na/H antiporter of E. coli.


Slightly soluble in aqueous base.


Synonyms: 4-carboxyphenylmercuric chloride; NSC 3286; p-chlormercuribenzoic acid; p-CMB

Formula: C₇H₅ClHgO₂
Melting Pt: >287 °C (dec.)
Storage Temperature: Low Temperature Freezer (–40°C)
CAS Number: 59-85-8
UN: 2025
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR