DBI-SALA® Protecta PRO™ Vest-Style Harnesses with Comfort Padding, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1191430 098-1191431
76111-328EA 103.17 USD
76111-328 76111-330
DBI-SALA® Protecta PRO™ Vest-Style Harnesses with Comfort Padding, ORS Nasco
Fall Protection Fall Harnesses
Durable yet lightweight construction translates to reduced worker fatigue and increased productivity.
  • Lightweight padding with soft edging wraps around the wearer for added comfort, security, and no-tangle donning
  • Back D-ring provides a safe connection point for a fall arrest system, and keeps users upright after a fall
  • Quick connect buckle leg and chest straps ensure fast, non-slipping connections
  • Parachute torso adjusters for a perfect fit
  • Lightweight and extremely strong polyester webbing provides ultimate durability and longevity
  • Built-in lanyard keepers to avoid trip hazards
  • Rip-stitch indicators allow users to inspect the harness at a glance, for prior damaging impact loads
  • Velcro® cover protects identification, inspection, and warning labels from the elements