Anti-Diglycyl-lysine Mouse Monoclonal Antibody [clone: GX41]

Supplier: Enzo Life Sciences
908-310-0100
76201-762EA 338 USD
76201-762
Anti-Diglycyl-lysine Mouse Monoclonal Antibody [clone: GX41]
Antibodies
Recommended Applications: IP, WB

Species reactivity: Species independent

Type: Primary
Antigen: Diglycyl-lysine
Clonality: Monoclonal
Clone: GX41
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Species independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR