Tiger, Ceramic Cutting Wheels, Weiler®
Supplier: Weiler
Tiger®
804-58302 804-58300 804-58305 804-58309
76184-212BX 229.17 USD
76184-212 76184-136 76184-224 76184-216
Tiger, Ceramic Cutting Wheels, Weiler®
Abrasives
Self-sharpening ceramic alumina grain delivers a superior combination of life and cut rate
  • Superior life and cut on metal
Use for cutting, edge beveling, edge softening, grinding, notching, scale removal, stock removal, weld blending, and weld removal applications.
Ideal for stainless steel and other hard-to-grind metals.