Protecta Cobra™ Rope Grabs, ORS Nasco
Supplier: ORS Nasco
098-AC202D
300075-692EA 196.95 USD
300075-692
Protecta Cobra™ Rope Grabs, ORS Nasco
Fall Protection Fall Arresters
Rope Grabs are designed for use on 5/8" polyester/polypropylene blended ropes
  • Cam locking system for safety
  • Detachable anywhere along the lifeline
They are designed to hold nylon or polyester rope on approved horizontal rope systems.
Certifications: Compliance with OSHA 1926 and ANSI Z359 Standards.