Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexahydropyridine Hexazane Pentamethyleneimine

571261-450ML 571261-200ML 571261-450ML 571261-200ML
BJ571261-450MLEA 241.81 USD
CABJ571261450M BJ571261-450ML CABJ571261200M BJ571261-200ML
Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™
Piperidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 107 °C
Melting Pt: -10 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 10.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00005979
CAS Number: 110-89-4
EINECS: 203-813-0
UN: 2401
ADR: 8,I

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Refractive index (n 20/D) 1.451 - 1.455
Water (Karl Fischer) Max. 0.3 %
Absorbance at 370 nm Max. 0.050
Absorbance at 290 nm Max. 1.000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR