Abacavir sulfate ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulphate((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulfate

M1612-5MG M1612-1MG
101969-320EA 738.85 USD
101969-320 101969-318
Abacavir sulfate ≥98% (by HPLC), MORpure™
Abacavir sulfate
Formula: C₂₈H₃₈N₁₂O₆S
MW: 670.74 g/mol
MDL Number: MFCD04112763
CAS Number: 188062-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR