Anti-MAPK13 Rabbit Polyclonal Antibody

Supplier: ProSci
62-960
76073-960EA 642.66 USD
76073-960
Anti-MAPK13 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50

Type: Primary
Antigen: MAPK13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 335-365
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR