Anti-LOH12CR1 Rabbit Polyclonal Antibody

Supplier: ProSci
62-054
76072-060EA 642.66 USD
76072-060
Anti-LOH12CR1 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: LOH12CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 167-196
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR